Learn More
Branson Ultrasonics™ Ultrasonic Cleaning Oxide Removers
Safely clean metals including stainless-steel, tin, and copper
$309.00
Specifications
Concentration 6 to 7%
Size 1 gal.
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Application) pH Range Product Type Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) pH Range Product Type Price Quantity & Availability  
15-335-46
View Documents
Branson Ultrasonics™
000955516
Rust, Metal Oxides 3 to 4 Oxide Remover
Case of 4 for $309.00
Add to cart
15-335-45
View Documents
Branson Ultrasonics™
000955316
Buffing compound, rouge, tripoli 6 to 7 Buffing Compound
Case of 4 for $309.00
Add to cart
Description
Description
  • Safely and effectively remove soils, rust, and oxides from a variety of materials
  • Solutions contain no phosphates and are biodegradable
  • Dilute with water to the recommended working concentration listed on ordering table
Specifications
Specifications
6 to 7%
1 gal.