Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Branson Ultrasonics™ Ultrasonic Industrial Cleaning Solutions

Catalog No. p-4880334
Click to view available options
Volume (English):
1 qt.
5 gal.

Safely clean metals including stainless-steel, tin, and copper

Industrial-strength.

  • Safely and effectively remove soils, rust, and oxides from a variety of materials
  • Solutions contain no phosphates and are biodegradable
  • Dilute with water to the recommended working concentration listed on ordering table

Specifications

pH 11 to 12
Concentration 8 to 10%
For Use With (Application) Cutting Oils, Metal Carbon
Volume (English) 1 qt.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.