Learn More
Ultraspec™ 2000 and 2001 Eyewear
SCI_ED
Adjustable side temple for a customized fit.
$9.23 - $12.16
Specifications
Certifications/Compliance Meet ANSI Z87.1-1989 standards for impact protection
Lens Tint Clear
Frame Color Clear
View More Specs
Products 2
Catalog Number Mfr. No. Lens Type Temple Style Price Quantity & Availability  
Catalog Number Mfr. No. Lens Type Temple Style Price Quantity & Availability  
S43187
SCI_ED
View Documents
Honeywell Safety Products
S0112C/EMD
Anti-fog Adjustable duoflex cushioned
Each for $12.16
Pack of 10 EA for $121.60
Add to cart
S43186
SCI_ED
View Documents
Honeywell Safety Products
S0250X/EMD
Scratch-resistant, Anti-fog Spatula
Each for $9.23
Pack of 10 EA for $92.27
Add to cart
Description
Description
  • Frameless lens features Ultradura™ hardcoat, scratch-resistant coating
  • S43187 has a larger lens to fit over most glasses and Duoflex™ comfort cushions at ends of the temples
Compliance
Meets ANSI™ Z87.1-1989 standards for impact protection.
Specifications
Specifications
Meet ANSI Z87.1-1989 standards for impact protection
Clear
Clear