Learn More
Youngstown Barrel & Drum Ultratech Foam Wall Model Ultra-Containment™ Berms
Low profile design for equipment maintenance and portable drum containment
$525.50 - $2900.00
Specifications
Type Ultra-Containment Berm
Model Foam wall
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Dimensions (L x W) Price Quantity & Availability  
19-110-008
View Documents
Youngstown Barrel & Drum
8461 UT
602 L 2.4 x 2.4 m
Each for $676.00
Add to cart
19-110-009
View Documents
Youngstown Barrel & Drum
8462 UT
1809 L 3.7 x 4.9 m
Each for $1,619.00
Add to cart
19-110-010
View Documents
Youngstown Barrel & Drum
8463 UT
3963 L 3.7 x 10.7 m
Each for $2,900.00
Add to cart
19-110-011
View Documents
Youngstown Barrel & Drum
8464 UT
5663 L 3.7 x 15.2 m
Each for $2,900.00
Add to cart
19-110-007
View Documents
Youngstown Barrel & Drum
8460 UT
223 L 1.2 x 1.8 m
Each for $525.50
Add to cart
Description
Description
  • Captures nuisance leaks and spills from vehicles, oily equipment, tanks and drums
  • Soft foam sidewall compresses for easy loading and off-loading
  • Withstands rigors of industrial and construction applications
  • Built-in handles help with transport and placement
Specifications
Specifications
Ultra-Containment Berm
Foam wall