Learn More
Youngstown Barrel & Drum Ultratech Rapid Rise Model Ultra-Containment™ Berms
Provides quick and easy spill containment for drums, IBCs, vehicles, frac tanks and other large containers
$1067.00 - $5780.00
Specifications
Type Ultra-Containment Berm
Model Rapid rise model
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Dimensions (L x W) Price Quantity & Availability  
19-110-003
View Documents
Youngstown Barrel & Drum
8630 UT
1018 L 1.8 x 1.8 m
Each for $1,067.00
Add to cart
19-110-004
View Documents
Youngstown Barrel & Drum
8631 UT
2831 L 3 x 3 m
Each for $1,734.00
Add to cart
19-110-005
View Documents
Youngstown Barrel & Drum
8632 UT
10,190 L 3.7 x 9.1 m
Each for $3,870.00
Add to cart
19-110-006
View Documents
Youngstown Barrel & Drum
8633 UT
16,989 L 3.7 x 15.2 m
Each for $5,780.00
Add to cart
Description
Description
  • During spill, foam ring around top perimeter of containment berm rises with level of spilled liquid
  • Sidewalls remain down during operations, loading and unloading
  • Built-in handles help with transport and placement
Specifications
Specifications
Ultra-Containment Berm
Rapid rise model