Learn More
BIOCHROM US ULTROSPEC 10 CELL DENSITY MET
Supplier:  BIOCHROM US 80211630
Encompass
Ultrospec 10 Cell Density Meter measures the density of cells in suspension at 600 nm. Small, portable, and easy to use, the instrument can measure growth rates of E. coli, other bacteria, and yeast (it is not recommended for use with mammalian cell lines). The hand-held device can be easily employed in incubation cabinets, under anaerobic conditions, and in other areas in the laboratory where cells are cultured. Spillages can be easily wiped from the smooth surface, and the cell compartment area can be flushed with, for example, formaldehyde or ethylene oxide to sterilize it. • Battery operated for use where cells are actually cultured. • Easy to use, easy to clean, easy to sterilize. • Download results to a PC or printer for convenient data storage and retrieval. • Small, portable, and dedicated cell density OD600 measurement instrument.
Catalog No. NC1144784