Learn More
Thermo Scientific™ Unassembled Convenience Kits: EPA Vials with Caps and Septa Available on GSA/VA Contract for Federal Government customers only.
Designed for discrete water sampling applications.
Manufacturer:  Thermo Scientific™ B7921B
 View more versions of this product
Catalog No. 03-377-37
  Check Availability
Add to cart
Description
Description
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
Each carton contains shrink-wrapped vials and separately packaged caps and natural-colored PTFE/clear Silicone septa in polybags.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
discrete water sampling applications
20mL
Amber 20mL vials, PTFE/Silicone septa and cap
Unassembled EPA Vial Kit
Amber
Amber; 20mL