Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC 1215, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1415800-43-9
C32H43N5O2
529.73
PQOOIERVZAXHBP-UHFFFAOYSA-N
2-phenylamino-1,4-phenylene bis 4-pyrrolidin-1-yl piperidin-1-yl methanone,2-phenylamino-1,4-bis 4-pyrrolidinyl piperidinyl benzamide,d0w9rr,unc hplc,2-phenylamino-1,4-4-pyrrolidinyl piperidinyl benzamide,2-phenylamino-1,4-bis 4-pyrrolidinyl-piperidinyl benzamide,n-phenyl-2,5-bis 4-pyrrolidin-1-yl piperidine-1-carbonyl aniline,n-phenyl-2,5-bis 4-pyrrolidin-1-yl piperidin-1-yl carbonyl aniline,3-anilino-4-4-pyrrolidin-1-ylpiperidine-1-carbonyl phenyl-4-pyrrolidin-1-ylpiperidin-1-yl methanone
57339144
This item is not returnable. View return policy
1415800-43-9
C32H43N5O2
529.73
PQOOIERVZAXHBP-UHFFFAOYSA-N
2-phenylamino-1,4-phenylene bis 4-pyrrolidin-1-yl piperidin-1-yl methanone,2-phenylamino-1,4-bis 4-pyrrolidinyl piperidinyl benzamide,d0w9rr,unc hplc,2-phenylamino-1,4-4-pyrrolidinyl piperidinyl benzamide,2-phenylamino-1,4-bis 4-pyrrolidinyl-piperidinyl benzamide,n-phenyl-2,5-bis 4-pyrrolidin-1-yl piperidine-1-carbonyl aniline,n-phenyl-2,5-bis 4-pyrrolidin-1-yl piperidin-1-yl carbonyl aniline,3-anilino-4-4-pyrrolidin-1-ylpiperidine-1-carbonyl phenyl-4-pyrrolidin-1-ylpiperidin-1-yl methanone
57339144

Potent inhibitor of L3MBTL3 Kme reader domain; cell permeable

Potent inhibitor of L3MBTL3 methyllysine (Kme) reader domain (IC50 = 40 nM; Kd = 120 nM). Displays >100-fold selectivity over a panel of histone methyltransferases, kinases, ion channels and 7-TM receptors. Disrupts subnuclear localization and foci formation of fluorescently-labeled L3MBTL3 in HEK293 cells.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 529.72
Percent Purity >99%
Chemical Name or Material UNC 1215

Recommended Storage : Store at -20°C