Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UNC 1215, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent inhibitor of L3MBTL3 methyllysine (Kme) reader domain (IC50 = 40 nM; Kd = 120 nM). Displays >100-fold selectivity over a panel of histone methyltransferases, kinases, ion channels and 7-TM receptors. Disrupts subnuclear localization and foci formation of fluorescently-labeled L3MBTL3 in HEK293 cells.
Specifications
Quantity | 10 mg |
Formula Weight | 529.72 |
Percent Purity | >99% |
Chemical Name or Material | UNC 1215 |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction