Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience UNC 2399
Biotinylated UNC 1999 (Cat.No. 4904)
Manufacturer: Tocris Bioscience 4930/1
This item is not returnable.
View return policy
Description
Biotinylated UNC 1999 (Cat.No. 4904, IC50 = 17 nM). Enriches lysine methyltransferase EZH2 from HEK293T cells.Specifications
1-Isopropyl-N-[(6-methyl-2-oxo-4-propyl-1,2-dihydropyridin-3-yl)methyl]-6-[6-[4-[41-oxo-45-[(3aS,4aS,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl]-4,7,10,13,16,19,22,25,28,31,34,37-dodecaoxa-40-azapentatetracontan-1oyl]piperazin-1-yl]pyridin-3-yl]-1H | |
C67H104N10O17S | |
1mg |
>95% | |
Observed MW: 1353.66 |
Safety and Handling
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title