Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Tocris Bioscience™ UNC 2400 Available on GSA/VA Contract for Federal Government customers only.
Negative control of UNC 1999 (Cat. No. 4904)
Manufacturer:  Tocris Bioscience™ 4905/10
Catalog No. 49-051-0
This item is not returnable. View return policy
Description
Description
Negative control of UNC 1999 . Exhibits 1000-fold lower potency than active analog (IC50 values are 62 and >200 μM for EZH1 and EZH2, respectively).
Active Analog also available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.