Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC 926 hydrochloride, Tocris Bioscience™

Catalog Number 451610
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
1782573-49-2
C16H22BrClN2O
373.719
DPPMBBHOXQOMJI-UHFFFAOYSA-N
unc 926 hydrochloride, unc-926 hydrochloride, 3-bromophenyl 4-1-prrolidinyl-1-piperidinyl methanone hydrochloride
90488947
(3-bromophenyl)-(4-pyrrolidin-1-ylpiperidin-1-yl)methanone;hydrochloride
C1CCN(C1)C2CCN(CC2)C(=O)C3=CC(=CC=C3)Br.Cl
This item is not returnable. View return policy
1782573-49-2
C16H22BrClN2O
373.719
DPPMBBHOXQOMJI-UHFFFAOYSA-N
unc 926 hydrochloride, unc-926 hydrochloride, 3-bromophenyl 4-1-prrolidinyl-1-piperidinyl methanone hydrochloride
90488947
(3-bromophenyl)-(4-pyrrolidin-1-ylpiperidin-1-yl)methanone;hydrochloride
C1CCN(C1)C2CCN(CC2)C(=O)C3=CC(=CC=C3)Br.Cl

L3MBTL1 domain inhibitor

Methyl-lysine (Kme) reader domain inhibitor; binds to the MBT domain of the L3MBTL1 protein (Kd = 3.9 μM). Selectively inhibits the L3MBTL13XMBT-H4K20me1 interaction in a peptide pull down assay.
Quantity 10 mg
Formula Weight 373.72
Percent Purity >99%
Chemical Name or Material UNC 926 hydrochloride
Recommended Storage Desiccate at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit