Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC 926 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1782573-49-2
C16H22BrClN2O
373.719
DPPMBBHOXQOMJI-UHFFFAOYSA-N
unc 926 hydrochloride,unc-926 hydrochloride,3-bromophenyl 4-1-prrolidinyl-1-piperidinyl methanone hydrochloride
90488947
(3-bromophenyl)-(4-pyrrolidin-1-ylpiperidin-1-yl)methanone;hydrochloride
C1CCN(C1)C2CCN(CC2)C(=O)C3=CC(=CC=C3)Br.Cl
This item is not returnable. View return policy
1782573-49-2
C16H22BrClN2O
373.719
DPPMBBHOXQOMJI-UHFFFAOYSA-N
unc 926 hydrochloride,unc-926 hydrochloride,3-bromophenyl 4-1-prrolidinyl-1-piperidinyl methanone hydrochloride
90488947
(3-bromophenyl)-(4-pyrrolidin-1-ylpiperidin-1-yl)methanone;hydrochloride
C1CCN(C1)C2CCN(CC2)C(=O)C3=CC(=CC=C3)Br.Cl

L3MBTL1 domain inhibitor

Methyl-lysine (Kme) reader domain inhibitor; binds to the MBT domain of the L3MBTL1 protein (Kd = 3.9 μM). Selectively inhibits the L3MBTL13XMBT-H4K20me1 interaction in a peptide pull down assay.
Quantity 50mg
Formula Weight 373.72
Percent Purity >99%
Chemical Name or Material UNC 926 hydrochloride

Recommended Storage : Desiccate at Room Temperature