Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UNC45A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 154791AP150UL
Description
UNC45A, also named as SMAP1, acts as co-chaperone for HSP90. UNC45A prevents the stimulation of HSP90AB1 ATPase activity by AHSA1. It is a positive factor in promoting PGR function in the cell. It is necessary for proper folding of myosin and necessary for normal cell proliferation, normal myotube formation and myosin accumulation during muscle cell development. UNC45A plays a role in erythropoiesis in stroma cells in the spleen.Specifications
UNC45A | |
Polyclonal | |
Unconjugated | |
UNC45A | |
GC UNC45, GCUNC 45, GCUNC45, IRO039700, Protein unc 45 homolog A, SMAP 1, SMAP1, unc 45 homolog A (C. elegans), Unc 45A, UNC45A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
101869, 308759, 55898 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q32PZ3, Q99KD5, Q9H3U1 | |
UNC45A | |
UNC45A Fusion Protein Ag7790 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction