Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UNC50 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569434
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
UNC50 is a protein coding gene.
Specifications
UNC50 | |
Polyclonal | |
Unconjugated | |
UNC50 | |
1110002A21Rik; geal-6 membrane-associated high-copy suppressor 1; GMH1; hGMH1; HSD23; HSD-23; hypothetical protein LOC513629; PDLs22; Periodontal ligament-specific protein 22; protein GMH1 homolog; Protein unc-50 homolog; UNC50; unc-50 homolog; unc-50 homolog (C. elegans); unc-50 inner nuclear membrane RNA binding protein; unc-50 related; unc-50 related protein (UNCL); unc-50-like protein; UNCL; uncoordinated-like protein; URP; zgc:56114 | |
Rabbit | |
Affinity chromatography | |
RUO | |
25972 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q53HI1 | |
UNC50 | |
synthetic peptide directed towards the N terminal region of human UNC50. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction