Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UNC84B Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | UNC84B |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB029303
|
Novus Biologicals
NBP293720100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB029304
|
Novus Biologicals
NBP29372020UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
UNC84B Polyclonal antibody specifically detects UNC84B in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
UNC84B | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.3), 50% glycerol | |
25777 | |
IgG | |
Affinity purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
FRIGG, KIAA0668, MGC133055, MGC133056, nuclear envelope protein, Protein unc-84 homolog B, Rab5-interacting protein, rab5IP, Sad1 and UNC84 domain containing 2, Sad1 unc-84 domain protein 2, Sad1/unc-84 protein-like 2, SUN domain-containing protein 2, unc-84 homolog B, unc-84 homolog B (C. elegans), UNC84B | |
Recombinant protein of human SUN2 | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title