Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UNC93A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA557377
Description
Immunogen sequence: QPIRDVQRES EGEKKSVPFW STLLSTFKLY RDKR Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 62%.
Unc93a, a human homolog of the C. elegans Unc93 gene, was initially identified by the sequencing of chromosome 6q27 at D6S193 as loss at this region correlated with sporadic ovarian cancer. Further analysis however, showed no tumor suppressor role for Unc93a. Unlike Unc93b, another homolog of the C. elegans Unc93 gene that is crucial for Toll-like receptor (TLR) signaling, the function of Unc93a is unknown. It is expressed in testis, small intestine, spleen, prostate and ovary.Specifications
UNC93A | |
Polyclonal | |
Unconjugated | |
UNC93A | |
dJ366N23.1; dJ366N23.1 (putative C. elegans UNC-93 (protein 1, C46F11.1) LIKE); dJ366N23.2; dJ366N23.2 (putative C. elegans UNC-93 (protein 1, C46F11.1) C-terminal LIKE); HmUnc-93A; Protein unc-93 homolog A; unc93 homolog A; unc-93 homolog A (C. elegans); UNC93A; Unc-93A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54346 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86WB7 | |
UNC93A | |
Recombinant protein corresponding to Human UNC93A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?
Provide Content Correction