Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNCX Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548137

Catalog No. PA548137

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Transcription factor involved in somitogenesis and neurogenesis. Required for the maintece and differentiation of particular elements of the axial skeleton. May act upstream of PAX9. Plays a role in controlling the development of connections of hypothalamic neurons to pituitary elements, allowing central neurons to reach the peripheral blood circulation and to deliver hormones for control of peripheral functions.
Specifications

Specifications

UNCX
Polyclonal
PBS with 0.09% sodium azide
A6NJT0
Homeobox protein unc-4 homolog, Homeobox protein Uncx41, UNCX, UNCX41
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
340260
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
UNCX
Liquid
UNCX
KLH conjugated synthetic peptide between 382-410 amino acids from the C-terminal region of human UNCX.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit