Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Undecanal, 97%

Click to view available options
Quantity:
25g
100g
112-44-7
C11H22O
170.296
MFCD00007016
KMPQYAYAQWNLME-UHFFFAOYSA-N
undecanaldehyde,n-undecanal,undecyl aldehyde,undecylic aldehyde,1-undecanal,undecylaldehyde,hendecanal,hendecanaldehyde,aldehyde c-11,n-undecyl aldehyde
8186
CHEBI:46202
undecanal
CCCCCCCCCCC=O
112-44-7
C11H22O
170.296
MFCD00007016
KMPQYAYAQWNLME-UHFFFAOYSA-N
undecanaldehyde,n-undecanal,undecyl aldehyde,undecylic aldehyde,1-undecanal,undecylaldehyde,hendecanal,hendecanaldehyde,aldehyde c-11,n-undecyl aldehyde
8186
CHEBI:46202
undecanal
CCCCCCCCCCC=O
Applications
Undecanal is used as a pharmaceutical intermediate.

Solubility
Soluble in dipropylene glycol, fixed oils, propylene glycol and water (14.27 mg/L at 25°C)

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, reducing agents and bases. It is sensitive to air.
Refractive Index 1.434
Density 0.827
Sensitivity Air Sensitive
Quantity 100g
Boiling Point 110°C to 112°C (12mmHg)
Melting Point -2°C
Flash Point 95°C (203°F)
Linear Formula CH3(CH2)9CHO
Beilstein 1753213
Solubility Information Soluble in dipropylene glycol,fixed oils,propylene glycol and water (14.27mg/L at 25°C)
Formula Weight 170.3
Percent Purity 97%
Chemical Name or Material Undecanal
Show More Show Less

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 203-972-6

RTECSNumber : YQ1500000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen