Alfa Aesar™ Undecanoic acid, 98%

Manufacturer: Alfa Aesar™ A1124430

Quantity 250g

 View more versions of this product

Catalog No. AAA1124430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 112-37-8
    Molecular Formula CH3(CH2)9CO2H
    Formula Weight 186.3
    MDL Number MFCD00002730
    Beilstein 1759287
    Chemical Name or Material Undecanoic acid

    Specifications

    Formula Weight 186.3
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula CH3(CH2)9CO2H
    CAS 112-37-8
    Health Hazard 1 Warning
    Melting Point 26° to 29°C
    Boiling Point 283° to 285°C
    Quantity 250g
    RTECS Number YQ2275000
    MDL Number MFCD00002730
    Beilstein 1759287
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point >110°C (230°F)
    Chemical Name or Material Undecanoic acid
    Assay 98%

    Description

    Applications
    Undecanoic acid is used as an unsaturated fatty acid. It is also used as intermediates of Liquid Crystals. It is used in organic synthesis, Catalytic agent, Petrochemical additive. It is also used in anti-fungal agent, perfumery chemicals.

    Solubility
    Insoluble in water.

    Notes
    Store at 4°C. Incompatible with bases, oxidizing agents, reducing agents. Keep container closed when not in use.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit