Learn More
GE Healthcare Uni-Core Manual Punch, 1.2mm Available on GSA/VA Contract for Federal Government customers only.
Achieve easy and cost-effective punching at the quality standards you expect. GE Healthcare Uni-Core Manual Punch product range provides manual punching for Whatman FTA, 903 and DMPK cards. Available in multiple diameters for maximum versatility.
$80.10 - $588.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
09-940-057 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
WB100074
25 Pieces Pack of 25 for $588.00
Add to cart
09-940-055 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
WB100028
4 Pieces Pack for $80.10
Add to cart
Description
Description
The Uni-Core range of manual punchers focuses on the products commonly needed for the preparation workflow of laboratory samples. Uni-Core punchers have a proprietary design with a sharp, stainless steel cutting edge. Using the recommended punch and cleaning procedure minimizes the risk of sample carryover between cards, while the companion cutting mat allows for clean sample cuts and extends the life of the cutting tip.
  • Designed for use with Whatman FTA Cards, FTA Elute Cards and EasiCollect devices
  • Diameters ranging from 1.0 to 6.0mm in plastic housing for sample punching versatility
  • Stainless steel cutting edges enable manual coring, retrieval and controlled ejection of the FTA punch
  • Disposable to minimize contamination and carryover between samples
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.