Learn More
Cytiva UNICORN Start 1.1, Distributed Electronically with License
UNICORN start is the control software designed for AKTA start.
Manufacturer:  Cytiva 29237234
Catalog No. 29-237-234
This product is not available for purchase online. For additional information, please contact your Fisher Scientific Technical Sales Representative.
Description
Description
UNICORN start is the control software designed for ÄKTA start.
  • Simple and flexible method creation using predefined templates and phases (steps)
  • Enables real-time monitoring of the run and an intuitive way to control the run
  • Evaluate, compare, and share results. Create and print PDF reports
  • Robust database storage with easy access to data, data security, and data integrity
UNICORN start software includes four different modules: System ControlMethod EditorEvaluation, and Administration, allowing you to design runs, operate the ÄKTA start instrument, and to evaluate and share results. While ÄKTA start can function as a stand-alone system, UNICORN start software maximizes the performance of the instrument.
Operating system:
  • Windows™ 7 Professional SP1 (US English version, 32-bit and 64-bit)
  • Windows™ 10 Professional (US English version, 32-bit and 64-bit) (Note: Computer and OS is not included with the purchase of UNICORN start)
License type: ESD with License 
Specifications
Specifications
Control Software
ESD with License
AKTA start