Learn More
Unifire Fog Machine Non-distribution product as customer accommodation.
Manufacturer:  Unifire UFZ1500
Catalog No. 19-120-3152
Add to cart
Description
Description
For users who need a lot of fog with a lot of control. Features the Z-2 remote with easy to read LED readout that allows the user to closely control the duration interval and volume of fog produced. Remote LED screen clearly displays the parameters in seconds. AC 120V/60Hz. 1500 watt heater. Output: 12,000 cu. ft./min. Weight: 30.86 lbs. (14.0kg) and measures 25.2 × 11.73 × 7.48 in. (64.0 × 29.8 × 19.0cm).
Specifications
Specifications
High performance smoke generating machine