Learn More
Unifire Pro Saw/Cut-Off Rescue Saw Non-distribution product as customer accommodation.
High-power, heavy-duty saw with reversible cutting arm is ideal for multiple rescue applications
Manufacturer:  Unifire PSCOS14
 View more versions of this product
Catalog No. 19-820-821
Please call customer service for current pricing. 1-800-766-7000
Specifications
Specifications
14 in. Blade
Cut off rescue saw, 14 in.
9.6kg
Large D-ring handle, smart start push-button decompression
21.3 lb.
Rescue applications that require cutting through materials including brick, steel or concrete