Learn More
Cole-Parmer Parker Hannifin Fitting, PFA, Straight, Compression Bulkhead Union, 3/8" OD  DFS Item
Manufacturer:  Cole-Parmer 3132084
Able to withstand higher pressures than barbed fittings, compression fittings are an excellent choice to connect rigid tubing systems.
Catalog No. 50-631-984