Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Cobalt Glass Plates
Cobalt Glass Plates used in flame tests to block orange color when performing sodium and potassium salt flame tests.
Supplier: United Scientific™ CGP3X3
Description
- Color: Dark blue
- Available in three different sizes
Specifications
Dark Blue | |
Cobalt Glass | |
7.6 x 7.6 cm (3 x 3 in.) | |
7.6 cm | |
7.6 cm |
Bunsen Burner | |
Non-sterile | |
3 in. | |
3 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction