Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Specific Gravity Cubes Products for Science Education

Ideal for specific gravity experiments.

$5.80 - $10.25

Specifications

For Use With (Application) To demonstrate the density of four different materials, specific gravity experiments
Product Type Density block
Includes Hook, specification sheet
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
S07031 Products for Science Education
View Documents
United Scientific Supplies
SGHC04BR
Brass Each for $10.25
Add to cart
 
S07032 Products for Science Education
View Documents
United Scientific Supplies
SGHC04AL
Aluminum Each for $5.80
Add to cart
 
Description

Description

  • Gravity cubes made of brass, aluminium and steel

Specifications

Specifications

To demonstrate the density of four different materials, specific gravity experiments
Density block
Hook, specification sheet
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit