Learn More
United Scientific Supplies Wide-Mouth Polypropylene Erlenmeyer Flasks Products for Science Education
For easy mixing and storage of solutions.
Manufacturer:  United Scientific Supplies FP0250
 View more versions of this product
Catalog No. S13080
Add to cart
Description
Description
  • Includes screw cap
  • Autoclavable
  • Graduated to show approximate volumes
  • Available in 125, 250 and 500mL capacities
Specifications
Specifications
Education
Screw
Polypropylene
8.4 oz.
250 mL
50 mL