Learn More
United Scientific Supplies Wide-Mouth Polypropylene Erlenmeyer Flasks Products for Science Education
For easy mixing and storage of solutions.
$32.75 - $44.00
Specifications
Material Polypropylene
Closure Type Screw
View More Specs
Products 2
Catalog Number Mfr. No. Graduations Price Quantity & Availability  
Catalog Number Mfr. No. Graduations Price Quantity & Availability  
S13079 Products for Science Education
View Documents
United Scientific Supplies
FP0125
25 mL
Pack of 12 for $32.75
Add to cart
S13080 Products for Science Education
View Documents
United Scientific Supplies
FP0250
50 mL
Pack of 12 for $44.00
Add to cart
Description
Description
  • Includes screw cap
  • Autoclavable
  • Graduated to show approximate volumes
  • Available in 125, 250 and 500mL capacities
Specifications
Specifications
Polypropylene
Screw