Learn More
Novus Biologicals Universal 3-Nitrotyrosine ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP266363
Description
The Universal 3-Nitrotyrosine ELISA Kit (Colorimetric) measures universal 3-Nitrotyrosine in serum, plasma and other biological fluids.Specifications
ELISA Kit (Colorimetric) | |
This kit recognizes 3-NT in samples. No significant cross-reactivity or interference between 3-NT and analogues was observed. | |
Storage is content dependent. | |
Universal | |
Biological Fluids, Plasma, Serum |
1.56 - 100 ng/mL | |
ELISA | |
3-Nitrotyrosine | |
1 Kit | |
0.94 ng/mL |