Learn More
Novus Biologicals Universal 5S-HETE ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP266722
Description
The Universal 5S-HETE ELISA Kit (Colorimetric) measures universal 5S-HETE in serum, plasma and other biological fluids.Specifications
ELISA Kit (Colorimetric) | |
This kit recognizes 5-HETE in samples. No significant cross-reactivity or interference between 5-HETE and analogues was observed. | |
Storage is content dependent. | |
Universal | |
Biological Fluids, Plasma, Serum |
0.16 - 10 ng/mL | |
ELISA | |
5S-HETE | |
1 Kit | |
0.09 ng/mL |