Learn More
Novus Biologicals™ Arachidonic Acid ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP266372
Catalog No. NBP266372
Add to Cart
Description
Description
The Universal Arachidonic Acid ELISA Kit (Colorimetric) measures universal Arachidonic Acid in serum, plasma and other biological fluids.
Specifications
Specifications
1.56 - 100 ng/mL
This kit recognizes AA in samples. No significant cross-reactivity or interference between AA and analogues was observed.
Biological Fluids, Plasma, Serum
Universal
Storage of components varies. See protocol for specific instructions.
0.94 ng/mL
ELISA Kit (Colorimetric)
ELISA
1 Kit
Arachidonic Acid
Product Suggestions
Product Suggestions