Learn More
Spectrum™ Universal Dialysis Tubing Closures (for all Membrane Types)
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Manufacturer:  Spectrum™ 142176
Catalog No. 08-700-246
Add to cart
Description
Description
  • Universal design with gentle sealing ridges, will not damage thicker walled tubing
  • Can be used with all membrane types: Standard RC, Biotech RC and Biotech CE
  • Easy to open and reseal
  • Color options for sample identification
  • Made of polyamide 6.6 (nylon)
  • Operating temperature: 0° to 90°C
  • Sterilizable by gamma irradiation or ethylene oxide (not autoclavable)
Specifications
Specifications
Non-sterile
Not Autoclavable
10 Pack