Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Universal Indicator, Reagents
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Specifications
Specifications
| Chemical Name or Material | Universal Indicator |
| CAS | 7732-18-5, 67-63-0, 67-63-0 |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 0.947 |
| pH | 6.8 to 7 |
| Quantity | 1 L |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction