Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Universal indicator Solution, pH 9 to 13, Honeywell™

Click to view available options
Quantity:
100mL
34722-90-2
C27H27Br2NaO5S
646.366
NMKFVGALBGZKGW-FKWCIMQXSA-M
102183223
sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate
CC1=C(C(=C(C=C1C(=C2C=C(C(=O)C(=C2C)Br)C(C)C)C3=CC=CC=C3S(=O)(=O)O)C(C)C)[O-])Br.[Na+]
34722-90-2
C27H27Br2NaO5S
646.366
NMKFVGALBGZKGW-FKWCIMQXSA-M
102183223
sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate
CC1=C(C(=C(C=C1C(=C2C=C(C(=O)C(=C2C)Br)C(C)C)C3=CC=CC=C3S(=O)(=O)O)C(C)C)[O-])Br.[Na+]

With color chart and directions for use, pH 9-13

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
UN Number UN1170
Quantity 100mL
Flash Point 22°C (71.6°F)
Chemical Name or Material Universal indicator solution

P210-P280-P305 + P351 + P338-P337 + P313-P403 + P235