Learn More
Novus Biologicals™ Universal Oxytocin ELISA Kit (Colorimetric)
This kit recognizes Universal OT in samples.
Manufacturer:  Novus Biologicals™ NBP280261
Catalog No. NB013332
Add to cart
Description
Description
No significant cross-reactivity or interference between Universal OT and analogues was observed.
Specifications
Specifications
15.625 to 1000 pg/mL
ELISA Kit (Colorimetric)
Oxytocin
5020
Universal
Storage of components varies. See protocol for specific instructions.
9.375 pg/mL
Serum, Plasma, Other Biological Fluids
ELISA
OXT
1 Kit