Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Specimen Clamp and Accessories

Get the most out of your Thermo Scientific™ microtome with this Thermo Scientific™ Specimen Clamp and Accessories.

Manufacturer:  Thermo Scientific™ 715060

 View more versions of this product

Catalog No. 715060


  Check Availability
Add to cart

Specifications

Description

Get the most out of your Thermo Scientific™ microtome with this Thermo Scientific™ Specimen Clamp and Accessories.

Segment Arc

  • For universal specimen holder with 360° rotation and standard accurate orientation of ±30°
  • Works in addition to specimen orientation in the X- and Y-axis
  • Settings via drive mechanism

Universal Specimen Holder

  • For flat and round specimens up to 8mm used with the segment arc

Standard Specimen Clamp

  • With fixed upper jaw ensuring greatest stability, orientation on X- and Y-axes, and rotation for accurate positioning
  • Maximum specimen size: 55 x 50mm
  • Silver model is for newer HM 325, HM 355S, and HM 340E models with serial numbers above 34000

Insert for Round Specimens

  • To be fitted in the standard specimen clamp
  • Tandem spring-loaded system ensures easy insertion of the specimen without tilting or squeezing
  • Optimal stability
  • Easy removal of the round specimen
  • When working with round specimen clamps, the standard specimen clamp must be equipped with a jaw light (Cat. No. 546080)

Jaw light used for weight reduction with inserts for round specimens

  • V-Insert for standard specimen clamps to assist in holding round specimens

  • V-Distance Piece for standard specimen clamps

  • Specimen Stages work with standard specimen clamp with V-insert

  • Universal Cassette Clamp with clamping jaws to be inserted in the microtome standard specimen clamp

    Foil Clamp

    • To be fitted in the standard specimen clamp
    • Spring-loaded jaws secure the specimen in position
    • A compensation slot in the jaw ensures perfect specimen clamping and constant section thicknesses
    • Orienting on Z-axis
    • Intermediate ring ensures alignment in X/Y zero-position
  • Description & Specifications

    Specifications

    Universal Specimen Holder
    Thermo Scientific™ Microtomes
    Universal Specimen Holder
    Universal specimen holder
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit