Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Universal Transport Medium

Hardy Diagnostics™ Universal Transport Medium is designed for collection, transport, maintenance and long term freeze storage of viruses, such as Influenza H1N1, Chlamydia, Mycoplasma and Ureaplasma specimens.

Manufacturer:  Hardy Diagnostics™ 302CHL

 View more versions of this product

Catalog No. 23-001-717

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

Description

Description

  • Has been used successfully for Rapid Antigen Testing, DFA, Viral Culture and for Molecular-based Assays.
  • Can be conveniently stored at room temperature prior to use.
  • Selection of tube sizes and collection applicators available.  Also available in bulk or as patient collection packs, which incorporate different combinations of Polyester swabs, as well as mini tip or regular flocked swabs (FLOQSwabs™) and media tubes to suit different work patterns and preferences.
  • Safe and reliable molded breakpoint swab - All swabs provided with UTM™ feature either a pre-scored breakpoint (polyester swabs) or a unique molded breakpoint (flocked swabs). Pre-scored and molded breakpoints feature allows for safe, reliable breaking of the swab into transport tube.
  • Shatterproof skirted tubes with distinctive internal conical shape enable centrifugation of samples and allows the tubes to stand upright on the laboratory bench.
  • Three glass beads in each tube facilitate the release and dispersion of patient sample material and virus particles from the swab during vortexing. 
Specifications

Specifications

Red
2 to 25°
UTM-RT Medium
300
3 mL
Polyester
16 x 100 mm
Light orange-red
Collection, transport, maintenance and long term freeze storage; Rapid Antigen Testing, DFA, Viral Culture and Molecular-based Assays.
Sterile
2 to 25°C
2 sterile wrapped polyester swabs
Screw Cap
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit