Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNKL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150001AP

Catalog No. 50-172-9900


Add to cart

Description

Description

May participate in a protein complex showing an E3 ligase activity regulated by RAC1. Ubiquitination is directed towards itself and possibly other substrates, such as SMARCD2/BAF60b. Intrinsic E3 ligase activity has not been proven.
Specifications

Specifications

UNKL
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5FWH2, Q9H9P5
C16orf28, KIAA0734, UNKL, ZC3H5L, ZC3HDC5L
Rabbit
IgG
150 μL
-20°C
Primary
302987, 64718, 74154
Western Blot
Unconjugated
UNKL
Liquid
UNKL
UNKL Fusion Protein Ag6990
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit