Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRSS55 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548266
Description
This gene encodes a member of a group of membrane-anchored chymotrypsin (S1)-like serine proteases. The enocoded protein is primarily expressed in the Leydig and Sertoli cells of the testis and may be involved in male fertility. Alternate splicing results in multiple transcript variants.
Specifications
PRSS55 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q6UWB4 | |
PRSS55 | |
KLH conjugated synthetic peptide between 289-318 amino acids from the C-terminal region of human UNQ9391. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
PRSS55 | |
Serine protease 55, 3421-, Testis serine protease 1, T-SP1, PRSS55, TSP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
203074 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction