Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Honeywell North™ Unsupported Butyl Gloves
Butyl has the highest permeation resistance to gas and water vapor, and is also highly resistant to ketones and esters.
$61.52
Specifications
Color Black
Exterior Finish Smooth
Glove Cuff Style Beaded
Material Butyl
Resistant To Gas, Water Vapor, Ketones, and Esters
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-085-590
View Documents
Honeywell North™
B174RGI/BAT/XS
X-Small Pair for $61.52
Case of 100 PR for $6,198.14
N/A
19-085-591
View Documents
Honeywell North™
B174RGI/BAT/S
Small Pair for $61.52
Case of 100 PR for $6,198.14
N/A
19-085-593
View Documents
Honeywell North™
B174RGI/BAT/M
Medium Pair for $61.52
Case of 100 PR for $6,198.14
N/A
19-085-594
View Documents
Honeywell North™
B174RGI/BAT/L
Large Pair for $61.52
Case of 100 PR for $6,198.14
N/A
Description
Description
  • For greater worker protection, especially when handling toxic substances
  • Flexible and sensitive, even at lower temperatures
  • Curved finger and hand design provides a better fit for enhanced comfort
Specifications
Specifications
Black
Beaded
Gas, Water Vapor, Ketones, and Esters
Unlined
ANSI/ISEA 105 (Abrasion Test, Cut Test)
Smooth
Butyl
14 in.
Unsupported
17 mil