Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

unsym-Dimethylethylenediamine, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
108-00-9
C4H12N2
88.15
MFCD00008175
DILRJUIACXKSQE-UHFFFAOYSA-N
n,n-dimethylethylenediamine,2-dimethylaminoethylamine,2-aminoethyl dimethylamine,1,2-ethanediamine, n,n-dimethyl,2-dimethylamino ethylamine,n,n-dimethyl-1,2-ethanediamine,n1,n1-dimethylethane-1,2-diamine,n,n-dimethyl-1,2-ethylenediamine,2-aminoethyldimethylamine,n,n-dimethylethanediamine
66053
N',N'-dimethylethane-1,2-diamine
CN(C)CCN
108-00-9
C4H12N2
88.15
MFCD00008175
DILRJUIACXKSQE-UHFFFAOYSA-N
n,n-dimethylethylenediamine,2-dimethylaminoethylamine,2-aminoethyl dimethylamine,1,2-ethanediamine, n,n-dimethyl,2-dimethylamino ethylamine,n,n-dimethyl-1,2-ethanediamine,n1,n1-dimethylethane-1,2-diamine,n,n-dimethyl-1,2-ethylenediamine,2-aminoethyldimethylamine,n,n-dimethylethanediamine
66053
N',N'-dimethylethane-1,2-diamine
CN(C)CCN
Melting Point -70°C
Boiling Point 105.5°C to 107°C
Flash Point 12°C
Refractive Index 1.4255 to 1.4275
Specific Gravity 0.8
Water 1% max.
Density 0.8000g/mL
Infrared Spectrum Authentic
Quantity 500g
Color Colorless to Yellow
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula (CH3)2NCH2CH2NH2
Beilstein 04, II, 690
Solubility Information Solubility in water: miscible with water
Formula Weight 88.15
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material unsym-Dimethylethylenediamine
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger