Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
uPA (PLAU) Mouse anti-Human, PE, Clone: UK98/6 (also known as UK 98/6; 98/6), BD Phrmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569971
Description
uPA (PLAU) Monoclonal antibody specifically detects uPA (PLAU) in Human samples. It is validated for Flow cytometry
Specifications
uPA (PLAU) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
5328 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Flow Cytometry | |
UK98/6 (also known as UK 98/6; 98/6) | |
PE | |
ATF; PLAU; U-plasminogen activator; UPA; u-PA; urokinase-type plasminogen activator | |
Purified Human Urokinase-type Plasminogen Activator | |
25 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction