Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ uPAR Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793225PLATES

 View more versions of this product

Catalog No. NB009684


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human uPAR.
Specifications

Specifications

HRP
Antibody Pairs
4.7 pg/mL
5329
RUO
uPAR
Cancer
Human
PLAUR
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.3 ng/mL is recommended.
Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human uPAR.
Sandwich ELISA
  • Standard: Each vial contains 17 ng of recombinant uPAR.
  • Capture Antibody: 0.3 mg/mL of mouse anti-uPAR monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.5 mg/mL mouse anti-uPAR monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Storage is content dependent.
    Mouse
    5 Plates
    4.69-300 pg/mL
    CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit