Learn More
uPAR Mouse anti-Human, Alexa Fluor 405, Clone: 62022, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB807V100UG
Catalog No. FAB807V100
Add to cart
This item is not returnable. View return policy
Description
Description
uPAR Monoclonal antibody specifically detects uPAR in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
uPAR
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q03405
PLAUR
Mouse myeloma cell line NS0-derived recombinant human uPAR Leu23-Arg303 Accession # Q03405
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human uPAR in direct ELISAs and Western blots. When used in a sandwich ELISA in combination with the biotinylated anti-human uPAR detection antibody (Catalog # BAF807), no significant cross-reactivity was observed with recombinant mouse uPAR.
Flow Cytometry
62022
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, PLAUR, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3
Mouse
IgG1
RUO
Primary
5329
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)