Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human uPAR Alexa Fluor 488-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB807G100UG
This item is not returnable.
View return policy
Description
uPAR Monoclonal specifically detects uPAR in Human samples. It is validated for Flow Cytometry.Specifications
uPAR | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q03405 | |
PLAUR | |
Mouse myeloma cell line NS0-derived recombinant human uPAR Leu23-Arg303 Accession # Q03405 | |
RUO | |
5329 | |
Human | |
IgG1 |
Flow Cytometry | |
62022 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, PLAUR, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
Mouse | |
100 μg | |
Primary | |
Detects human uPAR in direct ELISAs and Western blots. When used in a sandwich ELISA in combination with the biotinylated anti-human uPAR detection antibody (Catalog # BAF807), no significant cross-reactivity was observed with recombinant mouse uPAR. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction