Learn More
uPAR Rabbit anti-Human, Unconjugated, Clone: 2G10, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB122425UL
 View more versions of this product
Catalog No. ZRB122425MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
uPAR Monoclonal antibody specifically detects uPAR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Inhibition, Western Blotting.
Specifications
Specifications
uPAR
2G10
Unconjugated
Purified
PLAUR, MO3, UPAR
A recombinant fragment corresponding the first 277 amino acids from human Urokinase plasminogen activator surface receptor (uPAR).
Target Molecular Weight 36.98 kDa calculated; ∼45 kDa observed (non-reduced form). Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Inhibition Assays, Western Blot
300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_002650.1
Urokinase plasminogen activator surface receptor;CD87;U-PAR;Monocyte activation antigen Mo3
Rabbit
IgG
Protein A Purified
Signaling
Primary
Human