Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

uPAR Rabbit anti-Human, Unconjugated, Clone: 2G10, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen uPAR
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Inhibition Assays, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB122425MI
View Documents
MilliporeSigma
ZRB122425UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB12244XMI
View Documents
MilliporeSigma
ZRB12244X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

uPAR Monoclonal antibody specifically detects uPAR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Inhibition, Western Blotting.
Specifications

Specifications

uPAR
Unconjugated
Rabbit
Urokinase plasminogen activator surface receptor;CD87;U-PAR;Monocyte activation antigen Mo3
A recombinant fragment corresponding the first 277 amino acids from human Urokinase plasminogen activator surface receptor (uPAR).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Inhibition Assays, Western Blot
Purified
Human
PLAUR, MO3, UPAR
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit