Learn More
Upchurch Scientific™ Adapters for HPLC
Manufacturer:  Upchurch Scientific™ P135
 View more versions of this product
Catalog No. 05-700-74
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Adapter
0.08 in.
1000psi
White
1/4-28 flat bottom female to 5/16-24 female flat bottom
PEEK
4.12μL
Female to 5/16-24 female