Learn More
Idex Analytical Guard Column: Cartridge Holder Assembly
Guard columns and hardware to meet your system requirements
Manufacturer:  Idex C1000
Catalog No. 05-702-71
Add to cart
Description
Description
  • Extend the life of analytical columns
  • Save on column replacement costs
  • Include 10-32 coned threads for easy connections
  • Will hold to high pressures
  • Specially treated to prevent galling
Specifications
Specifications
Analytical Instrumentation
Cartridge Holder Assembly